Learn More
2-Bromo-9-fluorenone, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5596206
Description
2-Bromo-9-fluorenone, is used as organic intermediates functional materials. It is mainly used as best liquid crystal material. It is used as an OLED intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-9-fluorenone, is used as organic intermediates functional materials. It is mainly used as best liquid crystal material. It is used as an OLED intermediate.
Solubility
Solubility in other solvents, soluble in acetone.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
2-Bromo-9-fluorenone | |
146°C to 148°C | |
MFCD00010789 | |
2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 9h-fluoren-9-one, 2-bromo, 2-bromanylfluoren-9-one, acmc-209hjk, 2-bromo-9h-fluorenone, ksc318o8f | |
BrC1=CC=C2C3=CC=CC=C3C(=O)C2=C1 | |
259.10 | |
259.1 |
3096-56-8 | |
C13H7BrO | |
5 g | |
MTCARZDHUIEYMB-UHFFFAOYSA-N | |
2-bromofluoren-9-one | |
725831 | |
96% |
Chemical Identifiers
3096-56-8 | |
259.10 | |
MTCARZDHUIEYMB-UHFFFAOYSA-N | |
725831 | |
BrC1=CC=C2C3=CC=CC=C3C(=O)C2=C1 |
C13H7BrO | |
MFCD00010789 | |
2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 9h-fluoren-9-one, 2-bromo, 2-bromanylfluoren-9-one, acmc-209hjk, 2-bromo-9h-fluorenone, ksc318o8f | |
2-bromofluoren-9-one |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only