missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-9-fluorenone, 96%

Manufacturer:  Alfa Aesar™ H5596203

 View more versions of this product

Catalog No. AAH5596203



Description

Description

Applications
2-Bromo-9-fluorenone, is used as organic intermediates functional materials. It is mainly used as best liquid crystal material. It is used as an OLED intermediate.

Solubility
Solubility in other solvents, soluble in acetone.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

2-Bromo-9-fluorenone
1g
C13H7BrO
2-bromanylfluoren-9-one, 2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 2-bromo-9h-fluorenone, 9h-fluoren-9-one, 2-bromo, acmc-209hjk, ksc318o8f
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)Br
259.102
259.1
146°C to 148°C
3096-56-8
MFCD00010789
MTCARZDHUIEYMB-UHFFFAOYSA-N
2-bromofluoren-9-one
725831
96%

Chemical Identifiers

3096-56-8
259.102
MTCARZDHUIEYMB-UHFFFAOYSA-N
725831
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)Br
C13H7BrO
MFCD00010789
2-bromanylfluoren-9-one, 2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 2-bromo-9h-fluorenone, 9h-fluoren-9-one, 2-bromo, acmc-209hjk, ksc318o8f
2-bromofluoren-9-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents