Please login to your online account to display your discounted pricing

2-Bromo-6-(trifluoromethyl)pyridine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Bromo-6-(trifluoromethyl)pyridine
CAS 189278-27-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H3BrF3N
Formula Weight 226
MDL Number MFCD00153087
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB05802DA

 
fisher scientific company
SPB05802DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2-Bromo-6-(trifluoromethyl)pyridine
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Packaging Amber Glass Bottle
Molecular Formula C6H3BrF3N
MDL Number MFCD00153087
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
CAS 189278-27-1