Learn More
2-Bromo-6-pyridinemethanol, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5612306
Description
It is an active pharmaceutical ingredient and an pharmaceutical adjuvant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an active pharmaceutical ingredient and an pharmaceutical adjuvant.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents and bases. Store at room temperature.
Specifications
2-Bromo-6-pyridinemethanol | |
34°C to 39°C | |
>110°C (230°F) | |
MFCD04039884 | |
6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanol | |
XDDGKNRSCDEWBR-UHFFFAOYSA-N | |
(6-bromopyridin-2-yl)methanol | |
3517811 | |
96% |
33674-96-3 | |
246°C | |
C6H6BrNO | |
5 g | |
Slightly soluble in water. | |
OCC1=CC=CC(Br)=N1 | |
188.02 | |
188.02 |
Chemical Identifiers
33674-96-3 | |
188.02 | |
XDDGKNRSCDEWBR-UHFFFAOYSA-N | |
3517811 | |
OCC1=CC=CC(Br)=N1 |
C6H6BrNO | |
MFCD04039884 | |
6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanol | |
(6-bromopyridin-2-yl)methanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only