Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-(pentafluoroethyl)pyridine, 96%

Chemical Identifiers

Chemical Name or Material 2-Bromo-6-(pentafluoroethyl)pyridine
CAS 1026345-94-7
Formula Weight 276
MDL Number MFCD23382281
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66316MC

  SDS
alfa aesar™
H66316MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 276
Chemical Name or Material 2-Bromo-6-(pentafluoroethyl)pyridine
MDL Number MFCD23382281
Physical Form Viscous Liquid
CAS 1026345-94-7
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Yellow
Quantity 100mg
Assay 96%
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.