Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-methylpyridine, 98%

$92.36 - $382.72

Chemical Identifiers

Chemical Name or Material 2-Bromo-6-methylpyridine
CAS 5315-25-3
Molecular Formula C6H6BrN
Formula Weight 172.03
MDL Number MFCD00040743
Beilstein 107322
Synonym 6-Bromo-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2001806

  SDS
alfa aesar™
L2001806
5g Each for $92.36

AAL2001814

  SDS
alfa aesar™
L2001814
25g Each for $382.72
Description & Specifications

Specifications

Formula Weight 172.03
Chemical Name or Material 2-Bromo-6-methylpyridine
Assay 98%
Molecular Formula C6H6BrN
MDL Number MFCD00040743
CAS 5315-25-3
Beilstein 107322
Synonym 6-Bromo-2-picoline
Boiling Point 102° to 103°C (20mmHg)
Quantity 5g
Flash Point 97°C (206°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
6-Bromo-2-picoline is a halogenated pyridine derivative used is a building block in the preparation of nitrogen containing heterocyclic compounds. It may used in following synthesis 6,6′-dimethyl-2,2′-bipyridine, 6-methyl-2-pyridyl-2-pyridylmethanone, 2-methyl-6-(trimethylsilyl)-pyridine and N,N’-bis-(6-methylpyrid-2-yl)-(1R,2R)-1,2- diaminocyclohexane (cydiampy).

Solubility
Soluble in chloroform, ethyl aceate. Not miscible or difficult to mix with water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.