Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-methoxypyridine, 97%

$50.03 - $171.77

Chemical Identifiers

Chemical Name or Material 2-Bromo-6-methoxypyridine
CAS 40473-07-2
Molecular Formula C6H6BrNO
Formula Weight 188.02
MDL Number MFCD00088345
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3059314

  SDS
alfa aesar™
H3059314
25g Each for $171.77

AAH3059306

  SDS
alfa aesar™
H3059306
5g Each for $50.03
Description & Specifications

Specifications

Formula Weight 188.02
Chemical Name or Material 2-Bromo-6-methoxypyridine
Boiling Point 206°C
Quantity 25g
Flash Point 104°C (219°F)
Assay 97%
Molecular Formula C6H6BrNO
MDL Number MFCD00088345
CAS 40473-07-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.