missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-6-iodopyridine, TCI America™

$229.17 - $794.45

Chemical Identifiers

CAS 234111-08-1
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.894
MDL Number MFCD08059557
InChI Key KJOQMWRTNVBXEV-UHFFFAOYSA-N
Synonym pyridine, 2-bromo-6-iodo, 2-bromo-6-iodo-pyridine, 6-bromo-2-iodopyridine, pyridine,2-bromo-6-iodo
PubChem CID 11312130
IUPAC Name 2-bromo-6-iodopyridine
SMILES C1=CC(=NC(=C1)I)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B48151G View Documents TCI America
B48151G
1g Each for $229.17
 
B48155G View Documents TCI America
B48155G
5g Each for $794.45
 
Specifications

Chemical Identifiers

234111-08-1
283.894
KJOQMWRTNVBXEV-UHFFFAOYSA-N
11312130
C1=CC(=NC(=C1)I)Br
C5H3BrIN
MFCD08059557
pyridine, 2-bromo-6-iodo, 2-bromo-6-iodo-pyridine, 6-bromo-2-iodopyridine, pyridine,2-bromo-6-iodo
2-bromo-6-iodopyridine

Specifications

White-Yellow
1g
C5H3BrIN
pyridine, 2-bromo-6-iodo, 2-bromo-6-iodo-pyridine, 6-bromo-2-iodopyridine, pyridine,2-bromo-6-iodo
C1=CC(=NC(=C1)I)Br
283.894
283.89
≥98.0% (GC)
142°C
234111-08-1
MFCD08059557
KJOQMWRTNVBXEV-UHFFFAOYSA-N
2-bromo-6-iodopyridine
11312130
Crystal-Powder at 20°C
2-Bromo-6-iodopyridine
Safety and Handling

Safety and Handling

TSCA : No