Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-hydrazinopyridine, 98%

Chemical Identifiers

Chemical Name or Material 2-Bromo-6-hydrazinopyridine
CAS 26944-71-8
Formula Weight 188.03
MDL Number MFCD09743754
UN Number UN2811
Beilstein 386966
Synonym (6-Bromo-2-pyridyl)hydrazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6466403

  SDS
alfa aesar™
H6466403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6466406

  SDS
alfa aesar™
H6466406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6466414

  SDS
alfa aesar™
H6466414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.03
Melting Point 117° to 118°C
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Solid
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Assay 98%
Quantity 1g
Beilstein 386966
Color Pale brown
Health Hazard 2 GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
CAS 26944-71-8
MDL Number MFCD09743754
Chemical Name or Material 2-Bromo-6-hydrazinopyridine
UN Number UN2811
Health Hazard 1 Danger
Synonym (6-Bromo-2-pyridyl)hydrazine
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.