Learn More
2-Bromo-6-hydrazinopyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6466414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromo-6-hydrazinopyridine | |
117°C to 118°C | |
C5H6BrN3 | |
25 g | |
386966 | |
PQMFVUNERGGBPG-UHFFFAOYSA-N | |
(6-bromopyridin-2-yl)hydrazine | |
19392588 | |
98% |
26944-71-8 | |
Brown | |
MFCD09743754 | |
UN2811 | |
2-bromo-6-hydrazinylpyridine, 2-bromo-6-hydrazinopyridine, 2-6-bromopyridin-2-yl hydrazine, 6-bromo-pyridin-2-yl-hydrazine, 2-bromo-6-hydrazinopydine, 6-bromo-2-pyridyl hydrazine, 6-bromopyridin-2-yl hydrazine, 6-bromo-2-hydrazino pyridine, 2 1h-pyridinone, 6-bromo-, hydrazone, pubchem12987 | |
C1=CC(=NC(=C1)Br)NN | |
188.028 | |
188.03 | |
Solid |
Chemical Identifiers
26944-71-8 | |
188.028 | |
PQMFVUNERGGBPG-UHFFFAOYSA-N | |
19392588 | |
C1=CC(=NC(=C1)Br)NN |
C5H6BrN3 | |
MFCD09743754 | |
2-bromo-6-hydrazinylpyridine, 2-bromo-6-hydrazinopyridine, 2-6-bromopyridin-2-yl hydrazine, 6-bromo-pyridin-2-yl-hydrazine, 2-bromo-6-hydrazinopydine, 6-bromo-2-pyridyl hydrazine, 6-bromopyridin-2-yl hydrazine, 6-bromo-2-hydrazino pyridine, 2 1h-pyridinone, 6-bromo-, hydrazone, pubchem12987 | |
(6-bromopyridin-2-yl)hydrazine |
Safety and Handling
GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H301-H315-H318-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only