Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-fluorotoluene, 98%

$390.26 - $390.26

Chemical Identifiers

Chemical Name or Material 2-Bromo-6-fluorotoluene
CAS 1422-54-4
Molecular Formula C7H6BrF
Formula Weight 189.03
MDL Number MFCD00051438
Beilstein 2433658
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0751109

  SDS
alfa aesar™
L0751109
10g Each for $390.26
Description & Specifications

Specifications

Formula Weight 189.03
CAS 1422-54-4
Chemical Name or Material 2-Bromo-6-fluorotoluene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Assay 98%
Molecular Formula C7H6BrF
MDL Number MFCD00051438
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2433658
Boiling Point 180°C
Flash Point 76°C (168°F)
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.