missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-6-fluorobenzylamine hydrochloride, 96%, Thermo Scientific Chemicals
$315.32 - $1363.55
Chemical Identifiers
CAS | 1214330-90-1 |
---|---|
Molecular Formula | C7H8BrClFN |
Molecular Weight (g/mol) | 240.5 |
InChI Key | WEGQVRTUTQKDNY-UHFFFAOYSA-N |
Synonym | 2-bromo-6-fluorobenzylamine hcl, 2-bromo-6-fluorophenyl methanamine hydrochloride, 2-bromo-6-fluorophenyl methanamine;hydrochloride, 1-2-bromo-6-fluorophenyl methanamine hydrochloride |
PubChem CID | 118798502 |
IUPAC Name | (2-bromo-6-fluorophenyl)methanamine;hydrochloride |
SMILES | C1=CC(=C(C(=C1)Br)CN)F.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6208603
|
Thermo Scientific Chemicals
H6208603 |
1 g |
Each for $315.32
|
|
|||||
AAH6208606
|
Thermo Scientific Chemicals
H6208606 |
5 g |
Each for $1,363.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1214330-90-1 | |
240.5 | |
2-bromo-6-fluorobenzylamine hcl, 2-bromo-6-fluorophenyl methanamine hydrochloride, 2-bromo-6-fluorophenyl methanamine;hydrochloride, 1-2-bromo-6-fluorophenyl methanamine hydrochloride | |
(2-bromo-6-fluorophenyl)methanamine;hydrochloride |
C7H8BrClFN | |
WEGQVRTUTQKDNY-UHFFFAOYSA-N | |
118798502 | |
C1=CC(=C(C(=C1)Br)CN)F.Cl |
Specifications
1214330-90-1 | |
1 g | |
WEGQVRTUTQKDNY-UHFFFAOYSA-N | |
(2-bromo-6-fluorophenyl)methanamine;hydrochloride | |
118798502 | |
96% | |
2-Bromo-6-fluorobenzylamine hydrochloride |
C7H8BrClFN | |
2-bromo-6-fluorobenzylamine hcl, 2-bromo-6-fluorophenyl methanamine hydrochloride, 2-bromo-6-fluorophenyl methanamine;hydrochloride, 1-2-bromo-6-fluorophenyl methanamine hydrochloride | |
C1=CC(=C(C(=C1)Br)CN)F.Cl | |
240.5 | |
240.5 | |
Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only