missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-fluorobenzonitrile, 97%

Manufacturer:  Alfa Aesar™ H6149814

 View more versions of this product

Catalog No. AAH6149814



Specifications

Specifications

2-Bromo-6-fluorobenzonitrile
58°C to 62°C
79544-27-7
MFCD06797753
2-bromo-6-fluoro-benzonitrile
C1=CC(=C(C(=C1)Br)C#N)F
200.01
200.01
97%
White to Brown
25g
C7H3BrFN
UN3439
IELGUZKHALDFOO-UHFFFAOYSA-N
2-bromo-6-fluorobenzonitrile
2783139
Solid

Chemical Identifiers

79544-27-7
200.01
IELGUZKHALDFOO-UHFFFAOYSA-N
2783139
C1=CC(=C(C(=C1)Br)C#N)F
C7H3BrFN
MFCD06797753
2-bromo-6-fluoro-benzonitrile
2-bromo-6-fluorobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents