missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-fluorobenzoic acid, 97%

$90.96 - $324.65

Chemical Identifiers

CAS 2252-37-1
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD01569539
InChI Key MDAZJVAIZVUWDE-UHFFFAOYSA-N
Synonym 2-bromo-6-fluoro-benzoic acid
PubChem CID 302621
IUPAC Name 2-bromo-6-fluorobenzoic acid
SMILES C1=CC(=C(C(=C1)Br)C(=O)O)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6125103 View Documents Alfa Aesar™
H6125103
1g Each for $90.96
 
AAH6125106 View Documents Alfa Aesar™
H6125106
5g Each for $324.65
 
Specifications

Chemical Identifiers

2252-37-1
219.009
MDAZJVAIZVUWDE-UHFFFAOYSA-N
302621
C1=CC(=C(C(=C1)Br)C(=O)O)F
C7H4BrFO2
MFCD01569539
2-bromo-6-fluoro-benzoic acid
2-bromo-6-fluorobenzoic acid

Specifications

154°C to 158°C
2252-37-1
MFCD01569539
MDAZJVAIZVUWDE-UHFFFAOYSA-N
2-bromo-6-fluorobenzoic acid
302621
Solid
2-Bromo-6-fluorobenzoic acid
1g
C7H4BrFO2
2-bromo-6-fluoro-benzoic acid
C1=CC(=C(C(=C1)Br)C(=O)O)F
219.009
219.01
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures