missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-fluorobenzoic acid, 97%

Manufacturer:  Alfa Aesar™ H6125103

 View more versions of this product

Catalog No. AAH6125103



Specifications

Specifications

2-Bromo-6-fluorobenzoic acid
1g
C7H4BrFO2
2-bromo-6-fluoro-benzoic acid
C1=CC(=C(C(=C1)Br)C(=O)O)F
219.009
219.01
97%
154°C to 158°C
2252-37-1
MFCD01569539
MDAZJVAIZVUWDE-UHFFFAOYSA-N
2-bromo-6-fluorobenzoic acid
302621
Solid

Chemical Identifiers

2252-37-1
219.009
MDAZJVAIZVUWDE-UHFFFAOYSA-N
302621
C1=CC(=C(C(=C1)Br)C(=O)O)F
C7H4BrFO2
MFCD01569539
2-bromo-6-fluoro-benzoic acid
2-bromo-6-fluorobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents