Learn More
Alfa Aesar™ 2-Bromo-6-fluorobenzeneboronic acid, 98%
Manufacturer: Alfa Aesar™ H5307114
Specifications
2-Bromo-6-fluorobenzeneboronic acid | |
25g | |
C6H5BBrFO2 | |
2-bromo-6-fluorophenyl boronic acid, 2-bromo-6-fluorobenzeneboronic acid, 2-bromo-6-fluorophenylboronicacid, boronic acid, b-2-bromo-6-fluorophenyl, acmc-209ra3, 6-bromo-2-fluorophenylboronic acid, 2-bromanyl-6-fluoranyl-phenyl boronic acid | |
B(C1=C(C=CC=C1Br)F)(O)O | |
218.816 | |
218.82 |
142°C to 145°C | |
913835-80-0 | |
MFCD05664309 | |
MVSHYHSMIRBRGU-UHFFFAOYSA-N | |
(2-bromo-6-fluorophenyl)boronic acid | |
16217456 | |
98% |
Chemical Identifiers
913835-80-0 | |
218.816 | |
MVSHYHSMIRBRGU-UHFFFAOYSA-N | |
16217456 | |
B(C1=C(C=CC=C1Br)F)(O)O |
C6H5BBrFO2 | |
MFCD05664309 | |
2-bromo-6-fluorophenyl boronic acid, 2-bromo-6-fluorobenzeneboronic acid, 2-bromo-6-fluorophenylboronicacid, boronic acid, b-2-bromo-6-fluorophenyl, acmc-209ra3, 6-bromo-2-fluorophenylboronic acid, 2-bromanyl-6-fluoranyl-phenyl boronic acid | |
(2-bromo-6-fluorophenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning