missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-fluoroaniline, 98+%

$97.59 - $97.59

Chemical Identifiers

CAS 65896-11-9
Molecular Formula C6H5BrFN
Formula Weight 190.01
MDL Number MFCD01310982
UN Number UN2810
Beilstein 8541957
Chemical Name or Material 2-Bromo-6-fluoroaniline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2577406 SDS Alfa Aesar™
H2577406
5g Each for $97.59
AAH2577414 SDS Alfa Aesar™
H2577414
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

65896-11-9
190.01
UN2810
2-Bromo-6-fluoroaniline
C6H5BrFN
MFCD01310982
8541957

Specifications

65896-11-9
C6H5BrFN
MFCD01310982
8541957
≥98%
5g
190.01
UN2810
2-Bromo-6-fluoroaniline

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.