Please login to your online account to display your discounted pricing

2-Bromo-6-fluoroaniline, 95%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398880250

 View more versions of this product

Catalog No. AC398880250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 190.01
Quantity 25mL
Color Clear yellow to brown
Density 1.674g/mL
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Linear Formula BrC6H3(F)NH2
Refractive Index 1.5810 to 1.5830 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
MDL Number MFCD01310982
Molecular Formula C6H5BrFN
CAS 65896-11-9
Chemical Name or Material 2-Bromo-6-fluoroaniline
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Bromo-6-fluoroaniline
CAS 65896-11-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H5BrFN
Linear Formula BrC6H3(F)NH2
Formula Weight 190.01
MDL Number MFCD01310982
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents