missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Bromo-6'-fluoroacetophenone, 96%

$83.73 - $1,096.11

Chemical Identifiers

CAS 928715-37-1
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.037
InChI Key ZTLLHQUYOLPVAR-UHFFFAOYSA-N
Synonym 1-2-bromo-6-fluoro-phenyl ethanone, 1-2-bromo-6-fluorophenyl ethan-1-one, 1-2-bromo-6-fluorophenyl ethanone, 2'-bromo-6'-fluoroacetophenone, ethanone, 1-2-bromo-6-fluorophenyl
PubChem CID 66570680
IUPAC Name 1-(2-bromo-6-fluorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC=C1Br)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62361MD
View Documents
Alfa Aesar™
H62361MD
250mg Each for $83.73
 
AAH6236103
View Documents
Alfa Aesar™
H6236103
1g Each for $250.73
 
AAH6236106
View Documents
Alfa Aesar™
H6236106
5g Each for $1,096.11
 
Specifications

Chemical Identifiers

928715-37-1
217.037
1-2-bromo-6-fluoro-phenyl ethanone, 1-2-bromo-6-fluorophenyl ethan-1-one, 1-2-bromo-6-fluorophenyl ethanone, 2'-bromo-6'-fluoroacetophenone, ethanone, 1-2-bromo-6-fluorophenyl
1-(2-bromo-6-fluorophenyl)ethanone
C8H6BrFO
ZTLLHQUYOLPVAR-UHFFFAOYSA-N
66570680
CC(=O)C1=C(C=CC=C1Br)F

Specifications

250mg
C8H6BrFO
ZTLLHQUYOLPVAR-UHFFFAOYSA-N
1-(2-bromo-6-fluorophenyl)ethanone
66570680
Solid
2'-Bromo-6'-fluoroacetophenone
928715-37-1
1-2-bromo-6-fluoro-phenyl ethanone, 1-2-bromo-6-fluorophenyl ethan-1-one, 1-2-bromo-6-fluorophenyl ethanone, 2'-bromo-6'-fluoroacetophenone, ethanone, 1-2-bromo-6-fluorophenyl
CC(=O)C1=C(C=CC=C1Br)F
217.037
217.04
96%
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures