missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Bromo-6'-fluoroacetophenone, 96%

Manufacturer:  Alfa Aesar™ H6236106

 View more versions of this product

Catalog No. AAH6236106



Specifications

Specifications

2'-Bromo-6'-fluoroacetophenone
928715-37-1
1-2-bromo-6-fluoro-phenyl ethanone, 1-2-bromo-6-fluorophenyl ethan-1-one, 1-2-bromo-6-fluorophenyl ethanone, 2'-bromo-6'-fluoroacetophenone, ethanone, 1-2-bromo-6-fluorophenyl
CC(=O)C1=C(C=CC=C1Br)F
217.037
217.04
96%
5g
C8H6BrFO
ZTLLHQUYOLPVAR-UHFFFAOYSA-N
1-(2-bromo-6-fluorophenyl)ethanone
66570680
Solid

Chemical Identifiers

928715-37-1
217.037
1-2-bromo-6-fluoro-phenyl ethanone, 1-2-bromo-6-fluorophenyl ethan-1-one, 1-2-bromo-6-fluorophenyl ethanone, 2'-bromo-6'-fluoroacetophenone, ethanone, 1-2-bromo-6-fluorophenyl
1-(2-bromo-6-fluorophenyl)ethanone
C8H6BrFO
ZTLLHQUYOLPVAR-UHFFFAOYSA-N
66570680
CC(=O)C1=C(C=CC=C1Br)F
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents