missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-fluoro-4-methylpyridine, 98%

$72.47 - $862.04

Chemical Identifiers

CAS 180608-37-1
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD09839269
InChI Key CIDWWTCSJWQJQR-UHFFFAOYSA-N
Synonym 2-bromo-6-fluoro-4-picoline, 2-bromo4-methyl6-fluoropyridine, abbypharma ap-30-7592, 2-bromo-6-fluoro-4-methyl-pyridine, pyridine, 2-bromo-6-fluoro-4-methyl, pubchem6097, 2-bromo-6-fluoro-4-methyipyridine, pyridine,2-bromo-6-fluoro-4-methyl, 2-bromo-6-fluoro-4-methylpyridine;2-bromo-4-methyl-6-fluoropyridine
PubChem CID 29919366
IUPAC Name 2-bromo-6-fluoro-4-methylpyridine
SMILES CC1=CC(=NC(=C1)Br)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64449MD View Documents Alfa Aesar™
H64449MD
250mg Each for $72.47
 
AAH6444903 View Documents Alfa Aesar™
H6444903
1g Each for $215.89
 
AAH6444906 View Documents Alfa Aesar™
H6444906
5g Each for $862.04
 
Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

180608-37-1
190.015
CIDWWTCSJWQJQR-UHFFFAOYSA-N
29919366
CC1=CC(=NC(=C1)Br)F
C6H5BrFN
MFCD09839269
2-bromo-6-fluoro-4-picoline, 2-bromo4-methyl6-fluoropyridine, abbypharma ap-30-7592, 2-bromo-6-fluoro-4-methyl-pyridine, pyridine, 2-bromo-6-fluoro-4-methyl, pubchem6097, 2-bromo-6-fluoro-4-methyipyridine, pyridine,2-bromo-6-fluoro-4-methyl, 2-bromo-6-fluoro-4-methylpyridine;2-bromo-4-methyl-6-fluoropyridine
2-bromo-6-fluoro-4-methylpyridine

Specifications

Colorless to Yellow
180608-37-1
MFCD09839269
2-bromo-6-fluoro-4-picoline, 2-bromo4-methyl6-fluoropyridine, abbypharma ap-30-7592, 2-bromo-6-fluoro-4-methyl-pyridine, pyridine, 2-bromo-6-fluoro-4-methyl, pubchem6097, 2-bromo-6-fluoro-4-methyipyridine, pyridine,2-bromo-6-fluoro-4-methyl, 2-bromo-6-fluoro-4-methylpyridine;2-bromo-4-methyl-6-fluoropyridine
CC1=CC(=NC(=C1)Br)F
190.015
190.02
98%
250mg
C6H5BrFN
13574124
CIDWWTCSJWQJQR-UHFFFAOYSA-N
2-bromo-6-fluoro-4-methylpyridine
29919366
Liquid
2-Bromo-6-fluoro-4-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning