missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-6-fluoro-4-methylpyridine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6444903

 View more versions of this product

Catalog No. AAH6444903



Description

Description

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

2-Bromo-6-fluoro-4-methylpyridine
Colorless to Yellow
MFCD09839269
13574124
Slightly soluble in water.
CC1=CC(=NC(=C1)Br)F
190.015
190.02
Liquid
180608-37-1
C6H5BrFN
1 g
2-bromo-6-fluoro-4-picoline, 2-bromo4-methyl6-fluoropyridine, abbypharma ap-30-7592, 2-bromo-6-fluoro-4-methyl-pyridine, pyridine, 2-bromo-6-fluoro-4-methyl, pubchem6097, 2-bromo-6-fluoro-4-methyipyridine, pyridine,2-bromo-6-fluoro-4-methyl, 2-bromo-6-fluoro-4-methylpyridine;2-bromo-4-methyl-6-fluoropyridine
CIDWWTCSJWQJQR-UHFFFAOYSA-N
2-bromo-6-fluoro-4-methylpyridine
29919366
98%

Chemical Identifiers

180608-37-1
190.015
CIDWWTCSJWQJQR-UHFFFAOYSA-N
2-bromo-6-fluoro-4-methylpyridine
C6H5BrFN
MFCD09839269
29919366
CC1=CC(=NC(=C1)Br)F
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only