missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-ethoxynaphthalene, 98%

$104.34 - $358.97

Chemical Identifiers

CAS 66217-19-4
Molecular Formula C12H11BrO
Molecular Weight (g/mol) 251.123
MDL Number MFCD01568865
InChI Key FXVYIQCXQULCKL-UHFFFAOYSA-N
Synonym 2-bromo-6-ethoxy-naphthalene, 6-bromo-2-ethoxynaphthalene, 6-bromo-2-naphthyl ethyl ether, 6-ethoxy-2-bromonaphthalene, acmc-209nu2, naphthalene, 2-bromo-6-ethoxy, naphthalene,2-bromo-6-ethoxy
PubChem CID 236858
IUPAC Name 2-bromo-6-ethoxynaphthalene
SMILES CCOC1=CC2=C(C=C1)C=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50568MD
View Documents
Alfa Aesar™
H50568MD
250mg
Each for $104.34
 
AAH5056803
View Documents
Alfa Aesar™
H5056803
1g
Each for $358.97
 
Description

Description

Specifications

Chemical Identifiers

66217-19-4
251.123
FXVYIQCXQULCKL-UHFFFAOYSA-N
236858
CCOC1=CC2=C(C=C1)C=C(C=C2)Br
C12H11BrO
MFCD01568865
2-bromo-6-ethoxy-naphthalene, 6-bromo-2-ethoxynaphthalene, 6-bromo-2-naphthyl ethyl ether, 6-ethoxy-2-bromonaphthalene, acmc-209nu2, naphthalene, 2-bromo-6-ethoxy, naphthalene,2-bromo-6-ethoxy
2-bromo-6-ethoxynaphthalene

Specifications

250mg
C12H11BrO
2-bromo-6-ethoxy-naphthalene, 6-bromo-2-ethoxynaphthalene, 6-bromo-2-naphthyl ethyl ether, 6-ethoxy-2-bromonaphthalene, acmc-209nu2, naphthalene, 2-bromo-6-ethoxy, naphthalene,2-bromo-6-ethoxy
CCOC1=CC2=C(C=C1)C=C(C=C2)Br
251.123
251.12
2-Bromo-6-ethoxynaphthalene
66217-19-4
MFCD01568865
FXVYIQCXQULCKL-UHFFFAOYSA-N
2-bromo-6-ethoxynaphthalene
236858
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS