Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-chlorotoluene, 99%

$53.73 - $198.57

Chemical Identifiers

Chemical Name or Material 2-Bromo-6-chlorotoluene
CAS 62356-27-8
Molecular Formula C7H6BrCl
Formula Weight 205.48
MDL Number MFCD00061121
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2569706

  SDS
alfa aesar™
B2569706
5g Each for $53.73

AAB2569714

  SDS
alfa aesar™
B2569714
25g Each for $198.57
Description & Specifications

Specifications

Formula Weight 205.48
Molecular Formula C7H6BrCl
Boiling Point 120°C (40mmHg)
Quantity 5g
Chemical Name or Material 2-Bromo-6-chlorotoluene
Assay 99%
MDL Number MFCD00061121
CAS 62356-27-8
Flash Point 95°C (203°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.