Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-chloropyridine, 95%

$93.73 - $93.73

Chemical Identifiers

Chemical Name or Material 2-Bromo-6-chloropyridine
CAS 5140-72-7
Molecular Formula C5H3BrClN
Formula Weight 192.44
MDL Number MFCD00181262
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26961MD

  SDS
alfa aesar™
H26961MD
250mg Each for $93.73
Description & Specifications

Specifications

Formula Weight 192.44
CAS 5140-72-7
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 250mg
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Assay 95%
Chemical Name or Material 2-Bromo-6-chloropyridine
Molecular Formula C5H3BrClN
MDL Number MFCD00181262
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.