Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-chloro-4-(trifluoromethyl)aniline, 98%

Chemical Identifiers

Chemical Name or Material 2-Bromo-6-chloro-4-(trifluoromethyl)aniline
CAS 109919-26-8
Molecular Formula C7H4BrClF3N
Formula Weight 274.47
MDL Number MFCD00042180
UN Number UN2811
Synonym 4-Amino-3-bromo-5-chlorobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0923503

  SDS
alfa aesar™
L0923503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL09235MD

  SDS
alfa aesar™
L09235MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.47
Chemical Name or Material 2-Bromo-6-chloro-4-(trifluoromethyl)aniline
Assay 98%
Health Hazard 1 Danger
Molecular Formula C7H4BrClF3N
CAS 109919-26-8
MDL Number MFCD00042180
Synonym 4-Amino-3-bromo-5-chlorobenzotrifluoride
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
UN Number UN2811
Quantity 1g
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Flash Point >110°C (230°F)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 29° to 32°C
Boiling Point 71° to 72°C (0.25mmHg)
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.