Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-chloro-4-nitroaniline, 97%

Chemical Identifiers

Chemical Name or Material 2-Bromo-6-chloro-4-nitroaniline
CAS 99-29-6
Molecular Formula C6H4BrClN2O2
Formula Weight 251.47
MDL Number MFCD00014770
UN Number UN2811
Beilstein 2372771
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0219809

  SDS
alfa aesar™
L0219809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.47
Chemical Name or Material 2-Bromo-6-chloro-4-nitroaniline
MDL Number MFCD00014770
Molecular Formula C6H4BrClN2O2
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 99-29-6
Health Hazard 2 GHS H Statement
H301-H311-H331-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 10g
TSCA TSCA
Beilstein 2372771
Melting Point 173° to 177°C
UN Number UN2811
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.