Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-chloro-4-fluorophenol, 97%

$106.99 - $952.27

Chemical Identifiers

Chemical Name or Material 2-Bromo-6-chloro-4-fluorophenol
CAS 886499-83-8
Molecular Formula C6H3BrClFO
Formula Weight 225.45
MDL Number MFCD04115954
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2673403

  SDS
alfa aesar™
H2673403
1g Each for $106.99

AAH2673406

  SDS
alfa aesar™
H2673406
5g Each for $316.42

AAH2673414

  SDS
alfa aesar™
H2673414
25g Each for $952.27
Description & Specifications

Specifications

Formula Weight 225.45
Quantity 1g
Molecular Formula C6H3BrClFO
Health Hazard 1 Warning
Melting Point 38° to 40°C
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Chemical Name or Material 2-Bromo-6-chloro-4-fluorophenol
MDL Number MFCD04115954
CAS 886499-83-8
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2-Bromo-6-chloro-4-fluorophenol are employed in the preparation of resins, dyes, explosives, lubricants, and plastics. Also they are are endowed with antioxidant property and are used as antioxidants in many formulations such as pharmaceuticals, cosmetics, electrical transformer oil, solvents, and organic reactions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.