missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-6-chloro-4-fluoroaniline, 97%

Manufacturer:  Alfa Aesar™ A1539903

 View more versions of this product

Catalog No. AAA1539903



Specifications

Specifications

2-Bromo-6-chloro-4-fluoroaniline
54°C to 57°C
201849-14-1
MFCD00051707
benzenamine, 2-bromo-6-chloro-4-fluoro, pubchem2922, acmc-209f6h, attercop-chm at130703, 2-bromo-4-fluoro-6-chloroaniline, 2-bromo-6-chloro-4-fluoro-aniline, 2-bromo-6-chloro-4-fluorophenylamine, 6-bromo-2-chloro-4-fluorophenylamine, 2-bromo-6-chloro-4-fluoro-phenylamine, 2-bromo-6-chloro-4-fluoroaniline
C1=C(C=C(C(=C1Cl)N)Br)F
224.457
224.47
>110°C (230°F)
1g
C6H4BrClFN
UN2811
LIBGMUMMWYKJSC-UHFFFAOYSA-N
2-bromo-6-chloro-4-fluoroaniline
614401
97%

Chemical Identifiers

201849-14-1
224.457
LIBGMUMMWYKJSC-UHFFFAOYSA-N
614401
C1=C(C=C(C(=C1Cl)N)Br)F
C6H4BrClFN
MFCD00051707
benzenamine, 2-bromo-6-chloro-4-fluoro, pubchem2922, acmc-209f6h, attercop-chm at130703, 2-bromo-4-fluoro-6-chloroaniline, 2-bromo-6-chloro-4-fluoro-aniline, 2-bromo-6-chloro-4-fluorophenylamine, 6-bromo-2-chloro-4-fluorophenylamine, 2-bromo-6-chloro-4-fluoro-phenylamine, 2-bromo-6-chloro-4-fluoroaniline
2-bromo-6-chloro-4-fluoroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents