Learn More
Alfa Aesar™ 2-Bromo-6-chloro-4-fluoroaniline, 97%
Manufacturer: Alfa Aesar™ A1539906
Specifications
2-Bromo-6-chloro-4-fluoroaniline | |
54°C to 57°C | |
201849-14-1 | |
MFCD00051707 | |
benzenamine, 2-bromo-6-chloro-4-fluoro, pubchem2922, acmc-209f6h, attercop-chm at130703, 2-bromo-4-fluoro-6-chloroaniline, 2-bromo-6-chloro-4-fluoro-aniline, 2-bromo-6-chloro-4-fluorophenylamine, 6-bromo-2-chloro-4-fluorophenylamine, 2-bromo-6-chloro-4-fluoro-phenylamine, 2-bromo-6-chloro-4-fluoroaniline | |
C1=C(C=C(C(=C1Cl)N)Br)F | |
224.457 | |
224.47 |
>110°C (230°F) | |
5g | |
C6H4BrClFN | |
UN2811 | |
LIBGMUMMWYKJSC-UHFFFAOYSA-N | |
2-bromo-6-chloro-4-fluoroaniline | |
614401 | |
97% |
Chemical Identifiers
201849-14-1 | |
224.457 | |
LIBGMUMMWYKJSC-UHFFFAOYSA-N | |
614401 | |
C1=C(C=C(C(=C1Cl)N)Br)F |
C6H4BrClFN | |
MFCD00051707 | |
benzenamine, 2-bromo-6-chloro-4-fluoro, pubchem2922, acmc-209f6h, attercop-chm at130703, 2-bromo-4-fluoro-6-chloroaniline, 2-bromo-6-chloro-4-fluoro-aniline, 2-bromo-6-chloro-4-fluorophenylamine, 6-bromo-2-chloro-4-fluorophenylamine, 2-bromo-6-chloro-4-fluoro-phenylamine, 2-bromo-6-chloro-4-fluoroaniline | |
2-bromo-6-chloro-4-fluoroaniline |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.