missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-6-benzyloxypyridine, TCI America™

$181.81 - $627.92

Chemical Identifiers

CAS 117068-71-0
Molecular Formula C12H10BrNO
Molecular Weight (g/mol) 264.122
InChI Key WEMPEMXDSUATEK-UHFFFAOYSA-N
PubChem CID 817475
IUPAC Name 2-bromo-6-phenylmethoxypyridine
SMILES C1=CC=C(C=C1)COC2=NC(=CC=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B25765G View Documents TCI America
B25765G
5g Each for $181.81
 
B257625G View Documents TCI America
B257625G
25g Each for $627.92
 
Specifications

Chemical Identifiers

117068-71-0
264.122
817475
C1=CC=C(C=C1)COC2=NC(=CC=C2)Br
C12H10BrNO
WEMPEMXDSUATEK-UHFFFAOYSA-N
2-bromo-6-phenylmethoxypyridine

Specifications

White-Yellow
117068-71-0
WEMPEMXDSUATEK-UHFFFAOYSA-N
2-bromo-6-phenylmethoxypyridine
817475
Crystal-Lump at 20°C
2-Bromo-6-benzyloxypyridine
5g
C12H10BrNO
C1=CC=C(C=C1)COC2=NC(=CC=C2)Br
264.122
264.12
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator