missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+% PROMO, TCI America™

$92.20 - $459.58

Chemical Identifiers

CAS 651358-83-7
Molecular Formula C11H15BBrNO2
Molecular Weight (g/mol) 283.96
MDL Number MFCD06796177
InChI Key OAVRLCKBKDMGGW-UHFFFAOYSA-N
Synonym 2-6-bromo-2-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromo-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromopyridine-6-boronic acid pinacol ester, 6-bromo-2-pyridineboronic acid pinacol ester, 6-bromo-2-pyridylboronic acid pinacol ester, 6-bromopyridin-2-yl boronic acid pinacol ester, 6-bromopyridin-2-ylboronic acid pinacol ester, 6-bromopyridine-2-boronic acid pinacol ester, pyridine, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 12177444
IUPAC Name 2-bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC(Br)=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B50771GPM
View Documents Promotion Details
Tci America
B50771GPROMO
1 g
Each for $92.20
 
B50775GPM
View Documents Promotion Details
Tci America
B50775GPROMO
5 g
Each for $459.58
 
Description

Description

Specifications

Chemical Identifiers

651358-83-7
283.96
OAVRLCKBKDMGGW-UHFFFAOYSA-N
12177444
CC1(C)OB(OC1(C)C)C1=CC=CC(Br)=N1
C11H15BBrNO2
MFCD06796177
2-6-bromo-2-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromo-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromopyridine-6-boronic acid pinacol ester, 6-bromo-2-pyridineboronic acid pinacol ester, 6-bromo-2-pyridylboronic acid pinacol ester, 6-bromopyridin-2-yl boronic acid pinacol ester, 6-bromopyridin-2-ylboronic acid pinacol ester, 6-bromopyridine-2-boronic acid pinacol ester, pyridine, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
2-bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Specifications

651358-83-7
132°C
MFCD06796177
1 g
CC1(C)OB(OC1(C)C)C1=CC=CC(Br)=N1
283.96
283.96
≥98.0% (GC,T)
White
C11H15BBrNO2
2-6-bromo-2-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromo-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromopyridine-6-boronic acid pinacol ester, 6-bromo-2-pyridineboronic acid pinacol ester, 6-bromo-2-pyridylboronic acid pinacol ester, 6-bromopyridin-2-yl boronic acid pinacol ester, 6-bromopyridin-2-ylboronic acid pinacol ester, 6-bromopyridine-2-boronic acid pinacol ester, pyridine, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
OAVRLCKBKDMGGW-UHFFFAOYSA-N
2-bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
12177444
Crystal-Powder at 20°C
2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Safety and Handling

Safety and Handling

TSCA : No

SDS
Product Certifications