missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-(trimethylsilyl)thiophene, TCI America™

$305.56 - $1,063.36

Chemical Identifiers

CAS 18246-28-1
Molecular Formula C7H11BrSSi
Molecular Weight (g/mol) 235.214
InChI Key IXLYRDOGOXBECE-UHFFFAOYSA-N
Synonym (5-Bromo-2-thienyl)trimethylsilane
PubChem CID 13201448
IUPAC Name (5-bromothiophen-2-yl)-trimethylsilane
SMILES C[Si](C)(C)C1=CC=C(S1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B48551G View Documents TCI America
B48551G
1g Each for $305.56
 
B48555G View Documents TCI America
B48555G
5g Each for $1,063.36
 
Specifications

Chemical Identifiers

18246-28-1
235.214
(5-Bromo-2-thienyl)trimethylsilane
(5-bromothiophen-2-yl)-trimethylsilane
C7H11BrSSi
IXLYRDOGOXBECE-UHFFFAOYSA-N
13201448
C[Si](C)(C)C1=CC=C(S1)Br

Specifications

70°C
1g
C7H11BrSSi
IXLYRDOGOXBECE-UHFFFAOYSA-N
(5-bromothiophen-2-yl)-trimethylsilane
13201448
Clear Liquid at 20°C
2-Bromo-5-(trimethylsilyl)thiophene
Yellow
18246-28-1
(5-Bromo-2-thienyl)trimethylsilane
C[Si](C)(C)C1=CC=C(S1)Br
235.214
235.21
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No