Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-(trifluoromethyl)phenylacetonitrile, 97%

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-(trifluoromethyl)phenylacetonitrile
CAS 732306-26-2
Molecular Formula C9H5BrF3N
Formula Weight 264.04
MDL Number MFCD04973763
UN Number UN3439
Synonym 2-Bromo-5-(trifluoromethyl)benzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1959103

  SDS
alfa aesar™
L1959103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 264.04
Molecular Formula C9H5BrF3N
CAS 732306-26-2
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)phenylacetonitrile
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Assay 97%
MDL Number MFCD04973763
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
UN Number UN3439
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 60° to 62°C
Boiling Point 144° to 146°C (15mmHg)
Synonym 2-Bromo-5-(trifluoromethyl)benzyl cyanide
Quantity 1g
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.