Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-(trifluoromethyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-(trifluoromethyl)aniline
CAS 454-79-5
Molecular Formula C7H5BrF3N
Formula Weight 240.03
MDL Number MFCD00007637
UN Number UN2810
Beilstein 2094535
Synonym 3-Amino-4-bromobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1781003

  SDS
alfa aesar™
A1781003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1781006

  SDS
alfa aesar™
A1781006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1781014

  SDS
alfa aesar™
A1781014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.03
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 81° to 82°C (5mmHg)
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Assay 97%
UN Number UN2810
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)aniline
Beilstein 2094535
CAS 454-79-5
Molecular Formula C7H5BrF3N
Synonym 3-Amino-4-bromobenzotrifluoride
MDL Number MFCD00007637
Quantity 1g
Flash Point >110°C (230°F)

Applications
2-Bromo-5-(trifluoromethyl)aniline is used in the synthesis and biochemical evaluation of series of inhibitors of hepatitis C virus (HCV) NS3 protease.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.