Please login to your online account to display your discounted pricing

2-bromo-5-(trifluoromethyl)-1,3,4-thiadiazole, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC72310DE

 View more versions of this product

Catalog No. CC72310DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 233.01
Quantity 5g
Assay 97%
Packaging Amber Glass Bottle
Molecular Formula C3BrF3N2S
CAS 37461-61-3
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 2-bromo-5-(trifluoromethyl)-1, 3, 4-thiadiazole

Specifications

Chemical Name or Material 2-bromo-5-(trifluoromethyl)-1, 3, 4-thiadiazole
Assay 97%
Molecular Formula C3BrF3N2S
Formula Weight 233.01
Accessories

GHS Signal Word: Warning