Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-nitrotoluene, 98%

$56.81 - $56.81

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-nitrotoluene
CAS 7149-70-4
Molecular Formula C7H6BrNO2
Formula Weight 216.04
MDL Number MFCD00007281
Beilstein 1940342
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1545009

 
alfa aesar™
A1545009
10g Each for $56.81
Description & Specifications

Specifications

Formula Weight 216.04
Chemical Name or Material 2-Bromo-5-nitrotoluene
Melting Point 76° to 79°C
Quantity 10g
Assay 98%
Molecular Formula C7H6BrNO2
MDL Number MFCD00007281
CAS 7149-70-4
Beilstein 1940342
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.