missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-nitrotoluene, 98%

Manufacturer:  Alfa Aesar™ A1545018

 View more versions of this product

Catalog No. AAA1545018



Specifications

Specifications

2-Bromo-5-nitrotoluene
50g
C7H6BrNO2
1940342
HIMGPQVBNICCGL-UHFFFAOYSA-N
1-bromo-2-methyl-4-nitrobenzene
251672
98%
76°C to 79°C
7149-70-4
MFCD00007281
2-bromo-5-nitrotoluene, 4-bromo-3-methylnitrobenzene, benzene, 1-bromo-2-methyl-4-nitro, bromo 5--2-nitrotoluene, 2bromo-5-nitrotoluene, pubchem12553, acmc-209oiv, toluene, 2-bromo-5-nitro, ksc498a8d, methyl 2-bromo-5-nitrobenzene
CC1=C(C=CC(=C1)[N+](=O)[O-])Br
216.034
216.04

Chemical Identifiers

7149-70-4
216.034
HIMGPQVBNICCGL-UHFFFAOYSA-N
251672
CC1=C(C=CC(=C1)[N+](=O)[O-])Br
C7H6BrNO2
MFCD00007281
2-bromo-5-nitrotoluene, 4-bromo-3-methylnitrobenzene, benzene, 1-bromo-2-methyl-4-nitro, bromo 5--2-nitrotoluene, 2bromo-5-nitrotoluene, pubchem12553, acmc-209oiv, toluene, 2-bromo-5-nitro, ksc498a8d, methyl 2-bromo-5-nitrobenzene
1-bromo-2-methyl-4-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents