Please login to your online account to display your discounted pricing

2-Bromo-5-nitrothiazole, 98%, ACROS Organics™

$156.87 - $156.87

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-nitrothiazole
CAS 3034-48-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3HBrN2O2S
Formula Weight 209.02
MDL Number MFCD00005317
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC107180250

  SDS
thermo fisher scientific
AC107180250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC107180050

  SDS
thermo fisher scientific
107180050
Glass bottle 5g Each for $156.87
Description & Specifications

Specifications

Formula Weight 209.02
Physical Form Crystalline Powder
Assay ≥97.5%
Quantity 25g
Color Yellow to brown
Melting Point 84° to 88°C
CAS Max % 100.0
Chemical Name or Material 2-Bromo-5-nitrothiazole
Infrared Spectrum Authentic
Molecular Formula C3HBrN2O2S
CAS 3034-48-8
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Min % 97.5
MDL Number MFCD00005317
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.