missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-nitrothiazole, 98%

Manufacturer:  Alfa Aesar™ A1470103

 View more versions of this product

Catalog No. AAA1470103



Description

Description

Applications
2-Bromo-5-nitrothiazole is used as a building block. It is used as an intermediate to produce other chemicals.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-Bromo-5-nitrothiazole
1g
C3HBrN2O2S
4856
ANIJFZVZXZQFDH-UHFFFAOYSA-N
2-bromo-5-nitro-1,3-thiazole
18211
98%
85°C to 90°C
3034-48-8
MFCD00005317
2-bromo-5-nitrothiazole, thiazole, 2-bromo-5-nitro, 5-nitro-2-bromothiazole, pubchem11070, acmc-1ad6f, 2-bromo-5-nitro-thiazole, 4-27-00-00963 beilstein handbook reference, 2-bromo-5-nitrothiozole, 2-bromo-5-nitrothiazole, #
C1=C(SC(=N1)Br)[N+](=O)[O-]
209.017
209.03

Chemical Identifiers

3034-48-8
209.017
ANIJFZVZXZQFDH-UHFFFAOYSA-N
18211
C1=C(SC(=N1)Br)[N+](=O)[O-]
C3HBrN2O2S
MFCD00005317
2-bromo-5-nitrothiazole, thiazole, 2-bromo-5-nitro, 5-nitro-2-bromothiazole, pubchem11070, acmc-1ad6f, 2-bromo-5-nitro-thiazole, 4-27-00-00963 beilstein handbook reference, 2-bromo-5-nitrothiozole, 2-bromo-5-nitrothiazole, #
2-bromo-5-nitro-1,3-thiazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XJ2986000

TSCA : TSCA

Documents