Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-nitrobenzotrifluoride, 97%

$48.97 - $156.70

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-nitrobenzotrifluoride
CAS 367-67-9
Molecular Formula C7H3BrF3NO2
Formula Weight 270.01
MDL Number MFCD00014707
Beilstein 2460260
Synonym 1-Bromo-4-nitro-2-(trifluoromethyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1141606

  SDS
alfa aesar™
A1141606
5g Each for $48.97

AAA1141614

  SDS
alfa aesar™
A1141614
25g Each for $156.70
Description & Specifications

Specifications

Formula Weight 270.01
Health Hazard 1 Warning
Quantity 5g
Melting Point 41° to 45°C
Chemical Name or Material 2-Bromo-5-nitrobenzotrifluoride
Flash Point >110°C (230°F)
Beilstein 2460260
Molecular Formula C7H3BrF3NO2
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P337+P313
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Synonym 1-Bromo-4-nitro-2-(trifluoromethyl)benzene
CAS 367-67-9
MDL Number MFCD00014707
Assay 97%
Boiling Point 87° to 88°C (3mmHg)
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338-P337+P313
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.