missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-nitrobenzoic acid, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC342290010

 View more versions of this product

Catalog No. AC342290010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Bromo-5-nitrobenzoic acid
Glass bottle
180°C to 181°C
943-14-6
MFCD00134558
Solubility in water: insoluble
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Br
243025
Powder
Authentic
Beige to Brown
1g
C7H4BrNO4
2-bromo-5-nitro benzoic acid, 2-bromo-5-nitrobenzic acid, 2-bromo-5-nitrobenzoic acid, 2-bromo-5-nitrobenzoicacid, acmc-209rrc, benzoic acid, 2-bromo-5-nitro, ksc494m1r, paragos 440009, pubchem3821, timtec-bb sbb003179
UVFWYVCDRKRAJH-UHFFFAOYSA-N
246.02
246.02
≥96.0%

Chemical Identifiers

943-14-6
246.02
UVFWYVCDRKRAJH-UHFFFAOYSA-N
243025
C7H4BrNO4
MFCD00134558
2-bromo-5-nitro benzoic acid, 2-bromo-5-nitrobenzic acid, 2-bromo-5-nitrobenzoic acid, 2-bromo-5-nitrobenzoicacid, acmc-209rrc, benzoic acid, 2-bromo-5-nitro, ksc494m1r, paragos 440009, pubchem3821, timtec-bb sbb003179
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Br
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents