missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-5-nitroanisole 98.0+%, TCI America™
Supplier: TCI America B341825G
Specifications
2-Bromo-5-nitroanisole | |
104°C | |
MFCD00041250 | |
2-bromo-5-nitroanisole, 4-bromo-3-methoxynitrobenzene, 2-bromo-5-nitroanisol, benzene, 1-bromo-2-methoxy-4-nitro, 1-bromo-2-methoxy-4-nitro-benzene, zlchem 673, pubchem2536, 2-bromo 5-nitro anisole, 2-bromo-5-nitro-anisole, 2-bromo-5-nitro anisole | |
COC1=C(C=CC(=C1)[N+](=O)[O-])Br | |
232.033 | |
232.03 | |
Crystalline Powder |
77337-82-7 | |
C7H6BrNO3 | |
25 g | |
NTKADLOYTKVXQN-UHFFFAOYSA-N | |
1-bromo-2-methoxy-4-nitrobenzene | |
101293 | |
≥98.0% (GC) |
Chemical Identifiers
77337-82-7 | |
232.033 | |
NTKADLOYTKVXQN-UHFFFAOYSA-N | |
101293 | |
COC1=C(C=CC(=C1)[N+](=O)[O-])Br |
C7H6BrNO3 | |
MFCD00041250 | |
2-bromo-5-nitroanisole, 4-bromo-3-methoxynitrobenzene, 2-bromo-5-nitroanisol, benzene, 1-bromo-2-methoxy-4-nitro, 1-bromo-2-methoxy-4-nitro-benzene, zlchem 673, pubchem2536, 2-bromo 5-nitro anisole, 2-bromo-5-nitro-anisole, 2-bromo-5-nitro anisole | |
1-bromo-2-methoxy-4-nitrobenzene |
Safety and Handling
TSCA : No