Please login to your online account to display your discounted pricing

2-Bromo-5-nitroaniline, 98%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  334860010

 View more versions of this product

Catalog No. AC334860010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Fine Crystalline Needles or Crystalline Powder
Formula Weight 217.02
Quantity 1g
Color Dark yellow to khaki
Melting Point 138° to 142°C
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
CAS 10403-47-1
Molecular Formula C6H5BrN2O2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Chemical Name or Material 2-Bromo-5-nitroaniline
MDL Number MFCD00051578

Specifications

Chemical Name or Material 2-Bromo-5-nitroaniline
CAS 10403-47-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H5BrN2O2
Formula Weight 217.02
MDL Number MFCD00051578
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents