missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-n-octylthiophene, TCI America™

$330.00 - $1,145.86

Chemical Identifiers

CAS 172514-63-5
Molecular Formula C12H19BrS
Molecular Weight (g/mol) 275.248
InChI Key USLMRHXOAXBGCS-UHFFFAOYSA-N
PubChem CID 15440923
IUPAC Name 2-bromo-5-octylthiophene
SMILES CCCCCCCCC1=CC=C(S1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B37385G View Documents TCI America
B37385G
5g Each for $330.00
 
B373825G View Documents TCI America
B373825G
25g Each for $1,145.86
 
Specifications

Chemical Identifiers

172514-63-5
275.248
15440923
CCCCCCCCC1=CC=C(S1)Br
C12H19BrS
USLMRHXOAXBGCS-UHFFFAOYSA-N
2-bromo-5-octylthiophene

Specifications

112°C
5°C
172514-63-5
USLMRHXOAXBGCS-UHFFFAOYSA-N
2-bromo-5-octylthiophene
15440923
Clear Liquid at 20°C
2-Bromo-5-n-octylthiophene
Yellow
5g
C12H19BrS
CCCCCCCCC1=CC=C(S1)Br
275.248
275.25
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator