Learn More
2-Bromo-5-methylthiazole, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6395106
Description
2-Bromo-5-methylthiazole is a brominated thiazole compound for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-5-methylthiazole is a brominated thiazole compound for proteomics research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
2-Bromo-5-methylthiazole | |
C4H4BrNS | |
5 g | |
Slightly soluble in water. | |
CC1=CN=C(S1)Br | |
178.047 | |
178.05 |
41731-23-1 | |
MFCD08460610 | |
2-bromo-5-methylthiazole, 2-bromo-5-methyl-thiazole, thiazole, 2-bromo-5-methyl, pubchem20436, acmc-209jlg, 2-bromo-5-methyl thiazole, ksc493s9j | |
FJPZHYAYNAUKKA-UHFFFAOYSA-N | |
2-bromo-5-methyl-1,3-thiazole | |
21906106 | |
98% |
Chemical Identifiers
41731-23-1 | |
178.047 | |
FJPZHYAYNAUKKA-UHFFFAOYSA-N | |
21906106 | |
CC1=CN=C(S1)Br |
C4H4BrNS | |
MFCD08460610 | |
2-bromo-5-methylthiazole, 2-bromo-5-methyl-thiazole, thiazole, 2-bromo-5-methyl, pubchem20436, acmc-209jlg, 2-bromo-5-methyl thiazole, ksc493s9j | |
2-bromo-5-methyl-1,3-thiazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only