missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-5-methyl-4-phenylthiazole, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 412923-45-6 |
---|---|
Molecular Formula | C10H8BrNS |
Molecular Weight (g/mol) | 254.145 |
MDL Number | MFCD11220230 |
InChI Key | RNOQQISGUQUJMQ-UHFFFAOYSA-N |
Synonym | 2-bromo-5-methyl-4-phenylthiazole, thiazole, 2-bromo-5-methyl-4-phenyl, hydrochloride 1:1 |
PubChem CID | 49651940 |
IUPAC Name | 2-bromo-5-methyl-4-phenyl-1,3-thiazole |
SMILES | CC1=C(N=C(S1)Br)C2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5222603
|
Thermo Scientific Chemicals
H5222603 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
NotesStore in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
412923-45-6 | |
254.145 | |
RNOQQISGUQUJMQ-UHFFFAOYSA-N | |
49651940 | |
CC1=C(N=C(S1)Br)C2=CC=CC=C2 |
C10H8BrNS | |
MFCD11220230 | |
2-bromo-5-methyl-4-phenylthiazole, thiazole, 2-bromo-5-methyl-4-phenyl, hydrochloride 1:1 | |
2-bromo-5-methyl-4-phenyl-1,3-thiazole |
Specifications
412923-45-6 | |
MFCD11220230 | |
2-bromo-5-methyl-4-phenylthiazole, thiazole, 2-bromo-5-methyl-4-phenyl, hydrochloride 1:1 | |
CC1=C(N=C(S1)Br)C2=CC=CC=C2 | |
254.145 | |
254.14 | |
2-Bromo-5-methyl-4-phenylthiazole |
C10H8BrNS | |
1g | |
RNOQQISGUQUJMQ-UHFFFAOYSA-N | |
2-bromo-5-methyl-4-phenyl-1,3-thiazole | |
49651940 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures