missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-5-methoxytoluene, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 294030250
Specifications
2-Bromo-5-methoxytoluene | |
>110°C | |
Glass bottle | |
0.91 | |
Colorless to Yellow | |
27060-75-9 | |
100.0 | |
C8H9BrO | |
1-brom-4-methoxy-2-methylbenzol, 1-bromo-2-methyl-4-methoxybenzene, 2-bromo-5-methoxytoluene, 3-methyl-4-bromoanisole, 4-bromo-3-methyl-anisole, 4-bromo-3-methylanisole, acmc-1cg4z, benzene, 1-bromo-4-methoxy-2-methyl, pubchem3796, unii-okr329132j | |
CC1=C(C=CC(=C1)OC)Br | |
201.06 | |
201.06 | |
97% |
0.9100g/mL | |
Authentic | |
1.5600 to 1.5620 | |
111.0°C to 112.0°C (15.0 mmHg) | |
25g | |
96.0 | |
97% | |
MFCD00060687 | |
BLZNSXFQRKVFRP-UHFFFAOYSA-N | |
1-bromo-4-methoxy-2-methylbenzene | |
117915 | |
Liquid |
Chemical Identifiers
27060-75-9 | |
201.06 | |
BLZNSXFQRKVFRP-UHFFFAOYSA-N | |
117915 | |
CC1=C(C=CC(=C1)OC)Br |
C8H9BrO | |
MFCD00060687 | |
1-brom-4-methoxy-2-methylbenzol, 1-bromo-2-methyl-4-methoxybenzene, 2-bromo-5-methoxytoluene, 3-methyl-4-bromoanisole, 4-bromo-3-methyl-anisole, 4-bromo-3-methylanisole, acmc-1cg4z, benzene, 1-bromo-4-methoxy-2-methyl, pubchem3796, unii-okr329132j | |
1-bromo-4-methoxy-2-methylbenzene |