missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-methoxytoluene, 97%, Thermo Scientific™

Manufacturer:  Fisher Scientific Acros 294030050

 View more versions of this product

Catalog No. AC294030050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Bromo-5-methoxytoluene
>110°C
Glass bottle
0.91
Colorless to Yellow
27060-75-9
100.0
C8H9BrO
1-brom-4-methoxy-2-methylbenzol, 1-bromo-2-methyl-4-methoxybenzene, 2-bromo-5-methoxytoluene, 3-methyl-4-bromoanisole, 4-bromo-3-methyl-anisole, 4-bromo-3-methylanisole, acmc-1cg4z, benzene, 1-bromo-4-methoxy-2-methyl, pubchem3796, unii-okr329132j
CC1=C(C=CC(=C1)OC)Br
201.06
201.06
97%
0.9100g/mL
Authentic
1.5600 to 1.5620
111.0°C to 112.0°C (15.0 mmHg)
5g
96.0
97%
MFCD00060687
BLZNSXFQRKVFRP-UHFFFAOYSA-N
1-bromo-4-methoxy-2-methylbenzene
117915
Liquid

Chemical Identifiers

27060-75-9
201.06
BLZNSXFQRKVFRP-UHFFFAOYSA-N
117915
CC1=C(C=CC(=C1)OC)Br
C8H9BrO
MFCD00060687
1-brom-4-methoxy-2-methylbenzol, 1-bromo-2-methyl-4-methoxybenzene, 2-bromo-5-methoxytoluene, 3-methyl-4-bromoanisole, 4-bromo-3-methyl-anisole, 4-bromo-3-methylanisole, acmc-1cg4z, benzene, 1-bromo-4-methoxy-2-methyl, pubchem3796, unii-okr329132j
1-bromo-4-methoxy-2-methylbenzene
SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only