missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-methoxybenzyl bromide, 97%

Manufacturer:  Alfa Aesar™ H6496306

 View more versions of this product

Catalog No. AAH6496306



Description

Description

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Soluble in hot methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

2-Bromo-5-methoxybenzyl bromide
90°C to 94°C (literature)
19614-12-1
MFCD00078664
1945118
Soluble in hot methanol. Slightly soluble in water.
COC1=CC(=C(C=C1)Br)CBr
279.959
279.96
97%
White
5g
C8H8Br2O
UN3261
1-bromo-2-bromomethyl-4-methoxybenzene, 2-bromo-5-methoxy-benzyl bromide, 2-bromo-5-methoxybenzyl bromide, 2-bromo-5-methoxybenzylbromide, 4-bromo-3-bromomethyl anisole, 4-bromo-3-bromomethyl-1-methoxybenzene, 4-bromo3-bromomethylanisole, acmc-20a5x2, benzene, 1-bromo-2-bromomethyl-4-methoxy, pubchem18176
MURVUTUZSUEIGI-UHFFFAOYSA-N
1-bromo-2-(bromomethyl)-4-methoxybenzene
3525951
Solid

Chemical Identifiers

19614-12-1
279.959
MURVUTUZSUEIGI-UHFFFAOYSA-N
3525951
COC1=CC(=C(C=C1)Br)CBr
C8H8Br2O
MFCD00078664
1-bromo-2-bromomethyl-4-methoxybenzene, 2-bromo-5-methoxy-benzyl bromide, 2-bromo-5-methoxybenzyl bromide, 2-bromo-5-methoxybenzylbromide, 4-bromo-3-bromomethyl anisole, 4-bromo-3-bromomethyl-1-methoxybenzene, 4-bromo3-bromomethylanisole, acmc-20a5x2, benzene, 1-bromo-2-bromomethyl-4-methoxy, pubchem18176
1-bromo-2-(bromomethyl)-4-methoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents